5-Methoxynicotinaldehyde oxime

Supplier: Matrix Scientific

035205-250MG
101839-188EA 609.23 USD
101839-188
5-Methoxynicotinaldehyde oxime
5-Methoxynicotinaldehyde oxime

Hazard: Irritant

Formula: C₇H₈N₂O₂
MDL Number: MFCD11857644
CAS Number: 1087659-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR