2,3-Dimethoxy-6-(trimethylsilyl)pyridine

Supplier: Matrix Scientific

035203-250MG
101839-184EA 609.23 USD
101839-184
2,3-Dimethoxy-6-(trimethylsilyl)pyridine
2,3-Dimethoxy-6-(trimethylsilyl)pyridine

Hazard: Irritant

Formula: C₁₀H₁₇NO₂Si
MDL Number: MFCD11857642
CAS Number: 1131335-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR