6-Chloro-2,3-dimethoxypyridine

Supplier: Matrix Scientific

035202-250MG
101839-182EA 606.15 USD
101839-182
6-Chloro-2,3-dimethoxypyridine
6-Chloro-2,3-dimethoxypyridine

Hazard: Irritant

Formula: C₇H₈ClNO₂
MDL Number: MFCD11857641
CAS Number: 1087659-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR