2,6-Diiodo-3,5-dimethoxypyridine

Supplier: Matrix Scientific

035199-250MG
101839-176EA 606.15 USD
101839-176
2,6-Diiodo-3,5-dimethoxypyridine
2,6-Diiodo-3,5-dimethoxypyridine

Hazard: Irritant

Formula: C₇H₇I₂NO₂
MDL Number: MFCD11857638
CAS Number: 1131335-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR