2-Bromo-5-iodo-3-methoxypyridine

Supplier: Matrix Scientific

035194-250MG
101839-166EA 490.77 USD
101839-166
2-Bromo-5-iodo-3-methoxypyridine
2-Bromo-5-iodo-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₅BrINO
MDL Number: MFCD11857633
CAS Number: 1131335-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR