tert-Butyl-5-methoxypyridin-3-yl carbonate

Supplier: Matrix Scientific

035190-250MG
101839-158EA 606.15 USD
101839-158
tert-Butyl-5-methoxypyridin-3-yl carbonate
tert-Butyl-5-methoxypyridin-3-yl carbonate

Hazard: Irritant

Formula: C₁₁H₁₅NO₄
MW: 225.25 g/mol
MDL Number: MFCD11857629
CAS Number: 1131335-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR