5-Bromopyridin-3-yl-tert-butyl carbonate

Supplier: Matrix Scientific

035189-250MG
101839-156EA 604.62 USD
101839-156
5-Bromopyridin-3-yl-tert-butyl carbonate
5-Bromopyridin-3-yl-tert-butyl carbonate

Hazard: Irritant

Formula: C₁₀H₁₂BrNO₃
MW: 274.12 g/mol
MDL Number: MFCD11857628
CAS Number: 1087659-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR