3-Methoxy-5-(phenylcarbamoyl)isonicotinic acid
Supplier: Matrix Scientific
035187-250MG
101839-152EA
606.15
USD
101839-152
3-Methoxy-5-(phenylcarbamoyl)isonicotinic acid
3-Methoxy-5-(phenylcarbamoyl)isonicotinic acid
Hazard: Irritant
Formula:
C₁₄H₁₂N₂O₄ |
MDL Number:
MFCD11857626 CAS Number: 1087659-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...