3-Methoxy-5-(2-methyl-1,3-dioxolan-2-yl)pyridine

Supplier: Matrix Scientific

035185-250MG
101839-148EA 609.23 USD
101839-148
3-Methoxy-5-(2-methyl-1,3-dioxolan-2-yl)pyridine
3-Methoxy-5-(2-methyl-1,3-dioxolan-2-yl)pyridine

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD11857624
CAS Number: 1072933-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR