4-Chloro-5-methoxypyridin-3-amine

Supplier: Matrix Scientific

035183-250MG
101839-142EA 507.69 USD
101839-142
4-Chloro-5-methoxypyridin-3-amine
4-Chloro-5-methoxypyridin-3-amine

Hazard: Irritant

Formula: C₆H₇ClN₂O
MDL Number: MFCD11857622
CAS Number: 1105675-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR