3-Amino-5-methoxypyridin-4-ol dihydrochloride

Supplier: Matrix Scientific

035180-250MG
101839-136EA 609.23 USD
101839-136
3-Amino-5-methoxypyridin-4-ol dihydrochloride
3-Amino-5-methoxypyridin-4-ol dihydrochloride

Hazard: Irritant

Formula: C₆H₁₀Cl₂N₂O₂
MDL Number: MFCD11052860
CAS Number: 1105675-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR