3-Amino-5-methoxyisonicotinonitrile

Supplier: Matrix Scientific

035173-250MG
101839-122EA 606.15 USD
101839-122
3-Amino-5-methoxyisonicotinonitrile
3-Amino-5-methoxyisonicotinonitrile

Hazard: Irritant

Formula: C₇H₇N₃O
MDL Number: MFCD11052851
CAS Number: 1045855-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR