4-Iodo-5-methoxy-N-phenylnicotinamide

Supplier: Matrix Scientific

035172-250MG
101839-120EA 609.23 USD
101839-120
4-Iodo-5-methoxy-N-phenylnicotinamide
4-Iodo-5-methoxy-N-phenylnicotinamide

Hazard: Irritant

Formula: C₁₃H₁₁IN₂O₂
MDL Number: MFCD11857617
CAS Number: 1087659-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR