2-Bromo-6-iodo-3-methoxypyridine

035171-250MG
101839-118EA 211.13 USD
101839-118
2-Bromo-6-iodo-3-methoxypyridine
2-Bromo-6-iodo-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₅BrINO
MDL Number: MFCD09909341
CAS Number: 321535-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR