Methyl-3-(3-amino-5-methoxypyridin-4-yl)acrylate

Supplier: Matrix Scientific

035166-250MG
101839-108EA 606.15 USD
101839-108
Methyl-3-(3-amino-5-methoxypyridin-4-yl)acrylate
Methyl-3-(3-amino-5-methoxypyridin-4-yl)acrylate

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MDL Number: MFCD11052848
CAS Number: 1045858-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR