tert-Butyl-4-formyl-5-methoxypyridin-3-ylcarbamate

Supplier: Matrix Scientific

035160-250MG
101839-096EA 606.15 USD
101839-096
tert-Butyl-4-formyl-5-methoxypyridin-3-ylcarbamate
tert-Butyl-4-formyl-5-methoxypyridin-3-ylcarbamate

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₄
MW: 252.27 g/mol
MDL Number: MFCD11053566
CAS Number: 1049677-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR