tert-Butyl-5-methoxy-4-(trimethylsilyl)pyridin-3-ylcarbamate

Supplier: Matrix Scientific

035158-250MG
101839-092EA 606.15 USD
101839-092
tert-Butyl-5-methoxy-4-(trimethylsilyl)pyridin-3-ylcarbamate
tert-Butyl-5-methoxy-4-(trimethylsilyl)pyridin-3-ylcarbamate

Hazard: Irritant

Formula: C₁₄H₂₄N₂O₃Si
MW: 296.44 g/mol
MDL Number: MFCD11052842
CAS Number: 1045858-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR