tert-Butyl-4-chloro-5-methoxypyridin-3-ylcarbamate
Supplier: Matrix Scientific
035157-250MG
101839-090EA
606.15
USD
101839-090
tert-Butyl-4-chloro-5-methoxypyridin-3-ylcarbamate
tert-Butyl-4-chloro-5-methoxypyridin-3-ylcarbamate
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₂O₃ MW: 258.71 g/mol |
MDL Number:
MFCD11052841 CAS Number: 1045858-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...