3-(tert-Butoxycarbonylamino)-5-methoxyisonicotinic acid

Supplier: Matrix Scientific

035156-250MG
101839-088EA 492.31 USD
101839-088
3-(tert-Butoxycarbonylamino)-5-methoxyisonicotinic acid
3-(tert-Butoxycarbonylamino)-5-methoxyisonicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₅
MDL Number: MFCD11052840
CAS Number: 709666-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR