tert-Butyl-5-methoxy-4-methylpyridin-3-ylcarbamate
Supplier: Matrix Scientific
035153-250MG
101839-082EA
367.69
USD
101839-082
tert-Butyl-5-methoxy-4-methylpyridin-3-ylcarbamate
tert-Butyl-5-methoxy-4-methylpyridin-3-ylcarbamate
Hazard: Irritant
Formula:
C₁₂H₁₈N₂O₃ MW: 238.29 g/mol |
MDL Number:
MFCD11052838 CAS Number: 1045858-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...