5-Bromo-2-iodopyridin-3-ylacetate

Supplier: Matrix Scientific

035150-250MG
101839-076EA 364.62 USD
101839-076
5-Bromo-2-iodopyridin-3-ylacetate
5-Bromo-2-iodopyridin-3-ylacetate

Hazard: Irritant

Formula: C₇H₅BrINO₂
MW: 341.94 g/mol
MDL Number: MFCD11052836
CAS Number: 1045858-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR