5-Methoxynicotinonitrile

Supplier: Matrix Scientific

035147-250MG
101839-072EA 218.46 USD
101839-072
5-Methoxynicotinonitrile
5-Methoxynicotinonitrile

Hazard: Irritant

Formula: C₇H₆N₂O
MDL Number: MFCD09909444
CAS Number: 298204-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR