3-Iodo-5-nitropyridin-4-ol

Supplier: Matrix Scientific

035144-250MG
101839-068EA 606.15 USD
101839-068
3-Iodo-5-nitropyridin-4-ol
3-Iodo-5-nitropyridin-4-ol

Hazard: Irritant

Formula: C₅H₃IN₂O₃
MDL Number: MFCD11052833
CAS Number: 1072140-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR