2-Bromo-3-iodo-5-nitropyridine

Supplier: Matrix Scientific

035142-250MG
101839-064EA 364.62 USD
101839-064
2-Bromo-3-iodo-5-nitropyridine
2-Bromo-3-iodo-5-nitropyridine

Hazard: Irritant

Formula: C₅H₂BrIN₂O₂
MDL Number: MFCD09038021
CAS Number: 25391-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR