3-Iodo-5-nitropyridin-2-ol

Supplier: Matrix Scientific

035141-250MG
101839-062EA 367.69 USD
101839-062
3-Iodo-5-nitropyridin-2-ol
3-Iodo-5-nitropyridin-2-ol

Hazard: Irritant

Formula: C₅H₃IN₂O₃
MDL Number: MFCD06008303
CAS Number: 25391-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR