Methyl-3-(2-amino-4-methoxypyridin-3-yl)acrylate

Supplier: Matrix Scientific

035140-250MG
101839-060EA 606.15 USD
101839-060
Methyl-3-(2-amino-4-methoxypyridin-3-yl)acrylate
Methyl-3-(2-amino-4-methoxypyridin-3-yl)acrylate

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MDL Number: MFCD11052832
CAS Number: 1072139-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR