tert-Butyl-5-methoxypyridin-3-ylcarbamate

Supplier: Matrix Scientific

035138-250MG
101839-056EA 367.69 USD
101839-056
tert-Butyl-5-methoxypyridin-3-ylcarbamate
tert-Butyl-5-methoxypyridin-3-ylcarbamate

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃
MW: 224.26 g/mol
MDL Number: MFCD11052831
CAS Number: 342603-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR