2-Bromo-N-(1-phenylpropyl)acetamide

Supplier: Matrix Scientific

035120-500MG
101839-028EA 306.15 USD
101839-028
2-Bromo-N-(1-phenylpropyl)acetamide
2-Bromo-N-(1-phenylpropyl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₄BrNO
MDL Number: MFCD12026689
CAS Number: 1028257-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR