2-Bromo-N-(4-isopropoxybenzyl)acetamide

Supplier: Matrix Scientific

035119-500MG
101839-026EA 306.15 USD
101839-026
2-Bromo-N-(4-isopropoxybenzyl)acetamide
2-Bromo-N-(4-isopropoxybenzyl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD12026688
CAS Number: 947240-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR