4-[(2-Bromoacetyl)amino]-N-(1-phenylethyl)benzamide

Supplier: Matrix Scientific

035116-500MG
101839-020EA 306.15 USD
101839-020
4-[(2-Bromoacetyl)amino]-N-(1-phenylethyl)benzamide
4-[(2-Bromoacetyl)amino]-N-(1-phenylethyl)benzamide

Hazard: Irritant

Formula: C₁₇H₁₇BrN₂O₂
MW: 361.25 g/mol
MDL Number: MFCD12026685
CAS Number: 1138443-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR