N-Benzyl-4-[(2-bromoacetyl)amino]benzamide

Supplier: Matrix Scientific

035113-500MG
101839-014EA 306.15 USD
101839-014
N-Benzyl-4-[(2-bromoacetyl)amino]benzamide
N-Benzyl-4-[(2-bromoacetyl)amino]benzamide

Hazard: Irritant

Formula: C₁₆H₁₅BrN₂O₂
MDL Number: MFCD12026682
CAS Number: 1138443-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR