4-[(2-Bromoacetyl)amino]-N-(2-furylmethyl)benzamide

Supplier: Matrix Scientific

035109-500MG
101839-006EA 306.15 USD
101839-006
4-[(2-Bromoacetyl)amino]-N-(2-furylmethyl)benzamide
4-[(2-Bromoacetyl)amino]-N-(2-furylmethyl)benzamide

Hazard: Irritant

Formula: C₁₄H₁₃BrN₂O₃
MW: 337.18 g/mol
MDL Number: MFCD12026678
CAS Number: 1138443-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR