4-[(2-Bromoacetyl)amino]-N-ethyl-N-(1-naphthyl)benzamide

Supplier: Matrix Scientific

035106-500MG
101839-000EA 306.15 USD
101839-000
4-[(2-Bromoacetyl)amino]-N-ethyl-N-(1-naphthyl)benzamide
4-[(2-Bromoacetyl)amino]-N-ethyl-N-(1-naphthyl)benzamide

Hazard: Irritant

Formula: C₂₁H₁₉BrN₂O₂
MDL Number: MFCD12026676
CAS Number: 1138443-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR