3-[(2-Bromoacetyl)amino]-N-(1-phenylethyl)benzamide

Supplier: Matrix Scientific

035101-500MG
101838-990EA 306.15 USD
101838-990
3-[(2-Bromoacetyl)amino]-N-(1-phenylethyl)benzamide
3-[(2-Bromoacetyl)amino]-N-(1-phenylethyl)benzamide

Hazard: Irritant

Formula: C₁₇H₁₇BrN₂O₂
MW: 361.25 g/mol
MDL Number: MFCD12026671
CAS Number: 1138443-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR