3-[(2-Bromoacetyl)amino]-N,N-dipropylbenzamide

Supplier: Matrix Scientific

035098-500MG
101838-984EA 306.15 USD
101838-984
3-[(2-Bromoacetyl)amino]-N,N-dipropylbenzamide
3-[(2-Bromoacetyl)amino]-N,N-dipropylbenzamide

Hazard: Irritant

Formula: C₁₅H₂₁BrN₂O₂
MDL Number: MFCD12026668
CAS Number: 1138443-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR