2-Bromo-N-[3-(1-pyrrolidinylcarbonyl)phenyl]acetamide

Supplier: Matrix Scientific

035097-500MG
101838-982EA 306.15 USD
101838-982
2-Bromo-N-[3-(1-pyrrolidinylcarbonyl)phenyl]acetamide
2-Bromo-N-[3-(1-pyrrolidinylcarbonyl)phenyl]acetamide

Hazard: Irritant

Formula: C₁₃H₁₅BrN₂O₂
MW: 311.19 g/mol
MDL Number: MFCD12026667
CAS Number: 1138443-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR