3-[(2-Bromoacetyl)amino]benzamide

Supplier: Matrix Scientific

035095-500MG
101838-978EA 306.15 USD
101838-978
3-[(2-Bromoacetyl)amino]benzamide
3-[(2-Bromoacetyl)amino]benzamide

Hazard: Irritant

Formula: C₉H₉BrN₂O₂
MDL Number: MFCD12026665
CAS Number: 140215-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR