3-[(2-Bromoacetyl)amino]-N-(3-methoxypropyl)benzamide

Supplier: Matrix Scientific

035094-500MG
101838-976EA 306.15 USD
101838-976
3-[(2-Bromoacetyl)amino]-N-(3-methoxypropyl)benzamide
3-[(2-Bromoacetyl)amino]-N-(3-methoxypropyl)benzamide

Hazard: Irritant

Formula: C₁₃H₁₇BrN₂O₃
MW: 329.2 g/mol
MDL Number: MFCD12026664
CAS Number: 1138443-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR