N-[3-(1-Azepanylcarbonyl)phenyl]-2-bromoacetamide

Supplier: Matrix Scientific

035093-500MG
101838-974EA 306.15 USD
101838-974
N-[3-(1-Azepanylcarbonyl)phenyl]-2-bromoacetamide
N-[3-(1-Azepanylcarbonyl)phenyl]-2-bromoacetamide

Hazard: Irritant

Formula: C₁₅H₁₉BrN₂O₂
MDL Number: MFCD12026663
CAS Number: 1138443-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR