2-Bromo-N-(3-bromophenyl)acetamide

Supplier: Matrix Scientific

035089-500MG
101838-966EA 306.15 USD
101838-966
2-Bromo-N-(3-bromophenyl)acetamide
2-Bromo-N-(3-bromophenyl)acetamide

Hazard: Irritant

Formula: C₈H₇Br₂NO
MDL Number: MFCD02974391
CAS Number: 83276-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR