Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-[(2-Bromoacetyl)amino]-N-ethylbenzamide

Supplier: Matrix Scientific


035083-500MG
101838-954EA 260.23 USD InStock
101838-954
4-[(2-Bromoacetyl)amino]-N-ethylbenzamide
4-[(2-Bromoacetyl)amino]-N-ethylbenzamide
CAS: 1138443-16-9
MDL Number: MFCD12026657
Molecular Formula: C11H13BrN2O2
Molecular Weight: 285,15
Hazard: Irritant


Formula: C₁₁H₁₃BrN₂O₂
MW: 285,15 g/mol
MDL Number: MFCD12026657
CAS Number: 1138443-16-9
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty