3-[(2-Bromoacetyl)amino]-N,N-dimethylbenzamide

Supplier: Matrix Scientific

035077-500MG
101838-942EA 306.15 USD
101838-942
3-[(2-Bromoacetyl)amino]-N,N-dimethylbenzamide
3-[(2-Bromoacetyl)amino]-N,N-dimethylbenzamide

Hazard: Irritant

Formula: C₁₁H₁₃BrN₂O₂
MDL Number: MFCD12026652
CAS Number: 1138443-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR