3-[(2-Bromoacetyl)amino]-N-(3-chlorophenyl)benzamide

Supplier: Matrix Scientific

035072-500MG
101838-932EA 306.15 USD
101838-932
3-[(2-Bromoacetyl)amino]-N-(3-chlorophenyl)benzamide
3-[(2-Bromoacetyl)amino]-N-(3-chlorophenyl)benzamide

Hazard: Irritant

Formula: C₁₅H₁₂BrClN₂O₂
MW: 367.64 g/mol
MDL Number: MFCD12026647
CAS Number: 1138443-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR