2-Bromo-N-(4-methoxybenzyl)acetamide

Supplier: Matrix Scientific

035071-500MG
101838-930EA 306.15 USD
101838-930
2-Bromo-N-(4-methoxybenzyl)acetamide
2-Bromo-N-(4-methoxybenzyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂BrNO₂
MDL Number: MFCD12026646
CAS Number: 144581-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR