4-[(2-Bromoacetyl)amino]-N-methyl-N-phenylbenzamide

Supplier: Matrix Scientific

035070-500MG
101838-928EA 306.15 USD
101838-928
4-[(2-Bromoacetyl)amino]-N-methyl-N-phenylbenzamide
4-[(2-Bromoacetyl)amino]-N-methyl-N-phenylbenzamide

Hazard: Irritant

Formula: C₁₆H₁₅BrN₂O₂
MDL Number: MFCD12026645
CAS Number: 1138443-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR