4-[(2-Bromoacetyl)amino]-N-ethyl-N-phenylbenzamide

Supplier: Matrix Scientific

035069-500MG
101838-926EA 306.15 USD
101838-926
4-[(2-Bromoacetyl)amino]-N-ethyl-N-phenylbenzamide
4-[(2-Bromoacetyl)amino]-N-ethyl-N-phenylbenzamide

Hazard: Irritant

Formula: C₁₇H₁₇BrN₂O₂
MDL Number: MFCD12026644
CAS Number: 1138443-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR