3-[(2-Bromoacetyl)amino]-N-propylbenzamide

Supplier: Matrix Scientific

035066-500MG
101838-920EA 306.15 USD
101838-920
3-[(2-Bromoacetyl)amino]-N-propylbenzamide
3-[(2-Bromoacetyl)amino]-N-propylbenzamide

Hazard: Irritant

Formula: C₁₂H₁₅BrN₂O₂
MDL Number: MFCD12026641
CAS Number: 1138442-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR