2-Bromo-N-[3-(4-morpholinylcarbonyl)phenyl]acetamide

Supplier: Matrix Scientific

035059-500MG
101838-906EA 306.15 USD
101838-906
2-Bromo-N-[3-(4-morpholinylcarbonyl)phenyl]acetamide
2-Bromo-N-[3-(4-morpholinylcarbonyl)phenyl]acetamide

Hazard: Irritant

Formula: C₁₃H₁₅BrN₂O₃
MW: 327.19 g/mol
MDL Number: MFCD12026634
CAS Number: 926310-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR