4-[(2-Bromoacetyl)amino]-N-(3-methoxypropyl)benzamide

Supplier: Matrix Scientific

035058-500MG
101838-904EA 306.15 USD
101838-904
4-[(2-Bromoacetyl)amino]-N-(3-methoxypropyl)benzamide
4-[(2-Bromoacetyl)amino]-N-(3-methoxypropyl)benzamide

Hazard: Irritant

Formula: C₁₃H₁₇BrN₂O₃
MW: 329.2 g/mol
MDL Number: MFCD12026633
CAS Number: 1138442-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR