3-[(2-Bromoacetyl)amino]-N-(tetrahydro-2-furanylmethyl)benzamide

Supplier: Matrix Scientific

035054-500MG
101838-896EA 306.15 USD
101838-896
3-[(2-Bromoacetyl)amino]-N-(tetrahydro-2-furanylmethyl)benzamide
3-[(2-Bromoacetyl)amino]-N-(tetrahydro-2-furanylmethyl)benzamide

Hazard: Irritant

Formula: C₁₄H₁₇BrN₂O₃
MDL Number: MFCD12026631
CAS Number: 1138442-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR